Learn More
1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
1, 4-Dioxane, 99.5%, C4H8O2, CAS Number-123-91-1, diethylene dioxide, 1,4-dioxacyclohexane, 1,4-diethylene dioxide, dioxanne, dioxan, di ethylene oxide, diethylene ether, tetrahydro-p-dioxin, p-dioxane, dioxane, 1L, 0.002 % max., 19, 3, 101.0 deg.C, 100.0, 99.4, CHEBI:47032, Yellow to Brown
£31.66 - £192.96
Chemical Identifiers
CAS | 123-91-1 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.106 |
MDL Number | MFCD00006571 |
InChI Key | RYHBNJHYFVUHQT-UHFFFAOYSA-N |
Synonym | dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin |
PubChem CID | 31275 |
ChEBI | CHEBI:47032 |
IUPAC Name | 1,4-dioxane |
SMILES | C1COCCO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10762393
|
Thermo Scientific Acros
364341000 |
100 mL | AcroSeal™ Glass Bottle |
£31.66
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10352702
|
Thermo Scientific Acros
364340010 |
1 L | AcroSeal™ Glass Bottle |
£98.59
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10388533
|
Thermo Scientific Acros
364340025 |
2.5 L | AcroSeal™ Glass Bottle |
£192.96
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-91-1 | |
88.106 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
31275 | |
1,4-dioxane |
C4H8O2 | |
MFCD00006571 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
CHEBI:47032 | |
C1COCCO1 |
Specifications
12.0°C | |
99.4 | |
100.0 | |
101.0°C | |
Authentic | |
1.4205 to 1.4220 | |
1.034 | |
MFCD00006571 | |
1.32 mPa.s (20°C) | |
0.005% Max. | |
15, 3328 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents | |
C1COCCO1 | |
88.106 | |
CHEBI:47032 | |
99.5% | |
1, 4-Dioxane, Over Molecular Sieves |
123-91-1 | |
Yellow to Brown | |
1.0340g/mL | |
12°C | |
Liquid | |
99.5% | |
C4H8O2 | |
1165 | |
19, 3 | |
01,333 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
1,4-dioxane | |
31275 | |
88.11 | |
Extra Dry over Molecular Sieve |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
Emergency Overview
Flammable liquid and vapor. May form explosive peroxides. Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes
SIGNAL WORD: DANGER!
- 1,4-Dioxane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 1,4-Dioxane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.