missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur.

Product Code. p-217137 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic Drum
Unit Size:
1 L
10 L
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10687561

Brand: Thermo Scientific Acros 408825000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 12.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 1.0340g/mL
Boiling Point 101.0°C
Quantity 500 mL
Flash Point 12°C
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4205 to 1.4220
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 1.034
Titratable Acid 0.0016 meq/g max.
Viscosity 1.32 mPa.s (20°C)
Beilstein 19, 3
Water 0.05% max. (Coulometric)
Fieser 01,333
Merck Index 15, 3328
Formula Weight 88.11
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material 1, 4-Dioxane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Dioxane
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H350-May cause cancer.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-Dioxane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.