missing translation for 'onlineSavingsMsg'
Learn More
1,4-Dioxane, ACS reagent, Honeywell™ Product Code.: 15645070

1,4-Dioxane, ACS reagent, Honeywell™

Product Code. 15645070
2.5 L
Click to view available options
Quantity:
2.5 L
This item is not returnable. View return policy

Product Code. 15645070

Brand: Honeywell Riedel-de Haën 3604812.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 123-91-1
Density 1.034g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.22
Flash Point 12°C (53.6°F)
Ignition Point 356°F
Molecular Formula C4H8O2
MDL Number MFCD00006571
Vapor Pressure 27 mmHg (20°C), 40 mmHg (25°C)
UN Number UN1165
Beilstein 102551
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILES C1COCCO1
IUPAC Name 1,4-dioxane
Molecular Weight (g/mol) 88.106
PubChem CID 31275
ChEBI CHEBI:47032
Formula Weight 88.11g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material 1,4-Dioxane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Dioxane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Dioxane, ACS reagent, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.