missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, CHROMASOLV™ Plus, for HPLC, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15634550
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Narrow Mouth Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CHROMASOLV™ Plus, for HPLC,≥99.5%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 10°C to 12°C (literature)
Absorbance max. 0.01 at 295 nm, max. 0.10 at 270 nm, max. 0.20 at 250 nm, max. 0.50 at 235 nm, max. 0.70 at 220 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm
CAS Max % 100.0000%
Density 1.034 g/mL at 25 °C(lit.)
Boiling Point 100°C to 102°C
Quantity 100 mL
Flash Point 11°C
Packaging Glass Bottle
Linear Formula C4H8O2
UN Number UN1165
Beilstein 102551
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
Molecular Weight (g/mol) 88.106
Formula Weight 88.11
Percent Purity ≥99.5%
Grade HPLC
Chemical Name or Material 1,4-Dioxane, CHROMASOLV™ Plus, for HPLC, ≥99.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.