missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 6 to 8 (at 500g/L 20°C) |
Melting Point | 10°C to 12°C |
Color | Colorless |
Density | 1.034g/mL at 25°C |
Boiling Point | 100°C to 102°C |
Quantity | 4 x 2.5 L |
Explosion Limit | 0.22 |
Flash Point | 12°C (53.6°F) |
Ignition Point | 356°F |
Odor | Aromatic |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'