missing translation for 'onlineSavingsMsg'
Learn More

1,4-dioxaspiro[4.5]dec-8-ylamine, Thermo Scientific™

Product Code. 10235854
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 97096-16-7
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD04114556
InChI Key KDAFVGCPLFJMHY-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decan-8-amine, 1,4-dioxa-spiro 4.5 dec-8-ylamine, 8-amino-1,4-dioxaspiro 4,5 decane, 1,4-dioxaspiro 4.5 dec-8-ylamine, 1,4-dioxaspiro 4.5 decane-8-amine, 1,4-dioxa-spiro 4.5 dec-8-yl amine, 1,4-dioxa-spiro 4.5 dec-8-yl-amine
PubChem CID 14634315
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-amine
SMILES NC1CCC2(CC1)OCCO2

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material 1, 4-dioxaspiro[4.5]dec-8-ylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'