missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-dioxaspiro[4.5]dec-8-ylamine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Spécification
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 157.21 |
Percent Purity | 97% |
Chemical Name or Material | 1, 4-dioxaspiro[4.5]dec-8-ylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'