missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dithioerythritol, 98+%, Thermo Scientific Chemicals

Product Code. 11439402
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent for maintaining -SH groups in the reduced state, quantitatively reduces disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for maintaining -SH groups in the reduced state, quantitatively reduces disulfides.

Solubility
Soluble in water (15.4 mg/ml), and methanol.

Notes
Air Sensitive, store away form air. Keep the container tightly sealed. Store away from reducing agent. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6892-68-8
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00063750
InChI Key VHJLVAABSRFDPM-ZXZARUISSA-N
Synonym dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol
PubChem CID 439352
ChEBI CHEBI:17456
SMILES O[C@@H](CS)[C@H](O)CS

Specifications

Melting Point 82°C to 84°C
Odor Stench
Quantity 1 g
UN Number UN3335
Beilstein 1719756
IUPAC Name (2R,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Formula Weight 154.25
Percent Purity 99%
Chemical Name or Material 1,4-Dithioerythritol
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'