missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dithiothreitol, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15548774
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
POLYSTORMOR
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 3483-12-3
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S

Specifications

Flash Point 109.00°C
Packaging POLYSTORMOR
Quantity 5 g
Specific Gravity 1L = 1.00kg
Formula Weight 154.25
Grade Ultrapure Bioreagent
Chemical Name or Material 1,4-Dithiothreitol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'