missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Ethoxyphenyl)ethanol, 98%, Thermo Scientific™

Product Code. 15413857
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52067-36-4
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD08062382
InChI Key GKGQWBJLOYXULB-UHFFFAOYSA-N
Synonym 1-4-ethoxyphenyl ethan-1-ol, 1-4-ethoxyphenyl ethanol
PubChem CID 13891147
IUPAC Name 1-(4-ethoxyphenyl)ethanol
SMILES CCOC1=CC=C(C=C1)C(C)O

Specifications

Melting Point 46°C to 47°C
Quantity 5 g
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 1-(4-Ethoxyphenyl)ethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'