missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluoro-2-nitrophenyl)-4-methylpiperazine, 97%, Thermo Scientific™

Product Code. 15499758
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 432531-43-6
Molecular Formula C11H14FN3O2
Molecular Weight (g/mol) 239.25
MDL Number MFCD03144445
InChI Key SVXUJOMAWXJSIK-UHFFFAOYSA-N
Synonym 1-4-fluoro-2-nitrophenyl-4-methylpiperazine, cambridge id 6838238
PubChem CID 2915664
IUPAC Name 1-(4-fluoro-2-nitrophenyl)-4-methylpiperazine
SMILES CN1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 239.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)-4-methylpiperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'