missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluoro-2-nitrophenyl)piperidine, 97%, Thermo Scientific™

Product Code. 15449668
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 719-70-0
Molecular Formula C11H13FN2O2
Molecular Weight (g/mol) 224.235
MDL Number MFCD00798370
InChI Key YSBDMNNCMGUEAL-UHFFFAOYSA-N
Synonym 1-4-fluoro-2-nitrophenyl piperidine, n-4-fluoro-2-nitro-phenyl-piperidine, n-4-fluoro-2-nitro-phenyl piperidine, pubchem8030, acmc-1bbsb, 4-fluoro-2-nitrophenyl piperidine, piperidine,1-4-fluoro-2-nitrophenyl, 1-4-fluoranyl-2-nitro-phenyl piperidine
PubChem CID 2737434
IUPAC Name 1-(4-fluoro-2-nitrophenyl)piperidine
SMILES C1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 224.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)piperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.