missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluoro-3-methoxyphenyl)ethanol, 95%, Thermo Scientific™

Product Code. 15434807
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870849-56-2
Molecular Formula C9H11FO2
Molecular Weight (g/mol) 170.183
MDL Number MFCD08062372
InChI Key OGBGGCJKDXCBGP-UHFFFAOYSA-N
Synonym 1-4-fluoro-3-methoxyphenyl ethanol, 1-4-fluoro-3-methoxyphenyl ethan-1-ol, benzenemethanol,4-fluoro-3-methoxy-a-methyl
PubChem CID 24721520
IUPAC Name 1-(4-fluoro-3-methoxyphenyl)ethanol
SMILES CC(C1=CC(=C(C=C1)F)OC)O

Specifications

Quantity 5 g
Formula Weight 170.18
Percent Purity 95%
Chemical Name or Material 1-(4-Fluoro-3-methoxyphenyl)ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(4-Fluoro-3-methoxyphenyl)ethanol, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.