missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorophenyl)-1-methylethylamine, 97%, Thermo Scientific Chemicals

Product Code. 10411148
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17797-10-3
Molecular Formula C9H12FN
Molecular Weight (g/mol) 153.20
MDL Number MFCD07777176
InChI Key NYFSRXOWJXAHOO-UHFFFAOYSA-N
Synonym 2-4-fluorophenyl propan-2-amine, 1-4-fluorophenyl-1-methylethylamine, 1-4-fluorophenyl-1-methyl-ethylamine, 1-4-fluoro-phenyl-1-methyl-ethylamine, 2-4-fluorophenyl prop-2-ylamine, 1-methyl-1-4-fluorophenyl ethylamine, 4-fluorocumylamine, pubchem16914, 4-fluoro-alpha,alpha-dimethylbenzylamine, alpha,alpha-dimethyl-4-fluorobenzylamine
PubChem CID 13025665
IUPAC Name 2-(4-fluorophenyl)propan-2-amine
SMILES CC(C)(N)C1=CC=C(F)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 40.0°C (7.0 Mbar)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 153.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(4-Fluorophenyl)-1-methylethylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(4-Fluorophenyl)-1-methylethylamine, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.