missing translation for 'onlineSavingsMsg'
Learn More

1,4-Hexadiene, cis + trans, 99%

Product Code. 11490541
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11490541

Brand: Thermo Scientific Alfa Aesar B20060.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-45-0 | C6H10 | 82.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-45-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00009298
InChI Key PRBHEGAFLDMLAL-GQCTYLIASA-N
Synonym 1,4-hexadiene, trans-1,4-hexadiene, 1,4-hexadiene, 4e, 1,4-hexadiene,c&t, allylpropenyl, 1,4-hexadiene cis-trans, 1,4-hexa-1,4-diene, 1-allylpropene, 1,4-hexadiene,cis-trans, 1,4-trans-hexadiene
PubChem CID 5365552
IUPAC Name (4E)-hexa-1,4-diene
SMILES CC=CCC=C

Specifications

Density 0.71
Boiling Point 64°C to 66°C
Flash Point −26°C (−15°F)
Odor Characteristic
Refractive Index 1.411
Quantity 25 g
UN Number UN2458
Beilstein 1731889
Formula Weight 82.15
Percent Purity 99%
Chemical Name or Material 1,4-Hexadiene, cis + trans
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.