missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Methylphenyl)ethanol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.
Solubility
Slightly miscible with water.
Notes
Incompatible with oxidizing agents and strong reducing agents. Store in a cool place.
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.
Solubility
Slightly miscible with water.
Notes
Incompatible with oxidizing agents and strong reducing agents. Store in a cool place.
Specifications
Specifications
Density | 0.987 |
Boiling Point | 218°C to 220°C |
Flash Point | 107°C (225°F) |
Refractive Index | 1.523 |
Quantity | 1 g |
Beilstein | 1857562 |
Merck Index | 14,3237 |
Solubility Information | Slightly miscible with water. |
Formula Weight | 136.2 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-(4-Methylphenyl)ethanol, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction