missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methylphenyl)piperazine, 98%, Thermo Scientific Chemicals

£68.30 - £310.00

Chemical Identifiers

CAS 39593-08-3
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00040737
InChI Key ONEYFZXGNFNRJH-UHFFFAOYSA-N
Synonym 1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine
PubChem CID 83113
IUPAC Name 1-(4-methylphenyl)piperazine
SMILES CC1=CC=C(C=C1)N2CCNCC2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10712073
View Documents
Thermo Scientific Acros
370830050
5 g Glass bottle
£68.30
5g
Please to purchase this item. Need a web account? Register with us today!
10722073
View Documents
Thermo Scientific Acros
370830250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39593-08-3
176.26
ONEYFZXGNFNRJH-UHFFFAOYSA-N
83113
CC1=CC=C(C=C1)N2CCNCC2
C11H16N2
MFCD00040737
1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine
1-(4-methylphenyl)piperazine

Specifications

39593-08-3
100.0
White to Yellow
97.5% min. (GC)
MFCD00040737
ONEYFZXGNFNRJH-UHFFFAOYSA-N
1-(4-methylphenyl)piperazine
83113
98%
1-(4-Methylphenyl)piperazine
97.5
29°C to 34°C
Authentic
C11H16N2
1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine
CC1=CC=C(C=C1)N2CCNCC2
176.26
176.26
Low Melting Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 254-534-6

Product Identifier
  • 1-(4-Methylphenyl)piperazine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only