missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Naphthoquinone, 97+% (dry wt.), cont. up to 5% water, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.
Solubility
Soluble in alkali hydroxide solvents, sparingly soluble in petroleum ether and water. Freely soluble in carbondioxide, acetic acid, benzene and choloroform.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong reducing agents.
1,4-Naphthoquinone is used as a strong dienophile in Diels-Alder reaction. It is also used as a precursor to anthroquinone and 5-nitro-1,4-naphthalenedione, which finds application in the preparation of aminoanthroquinone and is also used as a dye precursor. It is a basic component of Vitamin K.
Solubility
Soluble in alkali hydroxide solvents, sparingly soluble in petroleum ether and water. Freely soluble in carbondioxide, acetic acid, benzene and choloroform.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong reducing agents.
Specifications
Specifications
Melting Point | 120°C to 124°C |
Density | 1.42 |
Flash Point | 142°C (288°F) |
Odor | Irritating |
Quantity | 100 g |
UN Number | UN2811 |
Beilstein | 878524 |
Merck Index | 14,6395 |
Solubility Information | Soluble in alkali hydroxide solvents,sparingly soluble in petroleum ether and water. Freely soluble in carbondioxide,acetic acid,benzene and choloroform. |
Formula Weight | 158.15 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'