missing translation for 'onlineSavingsMsg'
Learn More

1,4-Naphthoquinone, 99%, contains up to 6% water

Product Code. 10255500 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10255500

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 130-15-4 | C10H6O2 | 158.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-15-4, 7732-18-5
Molecular Formula C10H6O2
Molecular Weight (g/mol) 158.16
MDL Number MFCD00001676
InChI Key FRASJONUBLZVQX-UHFFFAOYSA-N
Synonym 1,4-naphthoquinone, 1,4-naphthalenedione, p-naphthoquinone, naphthoquinone, alpha-naphthoquinone, 1,4-naphthylquinone, usaf cy-10, 1,4-dihydronaphthalene-1,4-dione, 1,4-dihydro-1,4-diketonaphthalene, 1,4-naftochinon
PubChem CID 8530
ChEBI CHEBI:27418
IUPAC Name naphthalene-1,4-dione
SMILES C1=CC=C2C(=O)C=CC(=O)C2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.5°C
Color Khaki
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Beilstein 07, 724
Fieser 01,714
Merck Index 15, 6480
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkali hydroxide solutions,freely soluble in hot alcohol,ether,benzene,,chloroform,carbon bisulfide and acetic acid
Formula Weight 158.16
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 4-Naphthoquinone, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Naphthoquinone, contains up to 6% moisture
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.