Learn More
1,4-Pentadien-3-ol, 98%, stabilized, Thermo Scientific Chemicals
£98.59 - £312.56
Chemical Identifiers
CAS | 922-65-6 |
---|---|
Molecular Formula | C5H8O |
Molecular Weight (g/mol) | 84.12 |
MDL Number | MFCD00008633 |
InChI Key | ICMWSAALRSINTC-UHFFFAOYSA-N |
Synonym | 1,4-pentadien-3-ol, ccris 8963, divinylcarbinol, divinyl carbinol, acmc-20ap1x, 1,4-pentadien-3-ol, contains hydroquinone as stabilizer |
PubChem CID | 70204 |
IUPAC Name | penta-1,4-dien-3-ol |
SMILES | OC(C=C)C=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10411484
|
Thermo Scientific Acros
426240050 |
5 g | Glass bottle |
£98.59
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10623225
|
Thermo Scientific Acros
426240250 |
25 g | Glass bottle |
£312.56
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
922-65-6 | |
84.12 | |
ICMWSAALRSINTC-UHFFFAOYSA-N | |
70204 | |
OC(C=C)C=C |
C5H8O | |
MFCD00008633 | |
1,4-pentadien-3-ol, ccris 8963, divinylcarbinol, divinyl carbinol, acmc-20ap1x, 1,4-pentadien-3-ol, contains hydroquinone as stabilizer | |
penta-1,4-dien-3-ol |
Specifications
922-65-6 | |
100.0 | |
0.8650g/mL | |
30°C | |
97.5% min. (GC) | |
(CH2CH)2CHOH | |
MFCD00008633 | |
1,4-pentadien-3-ol, ccris 8963, divinylcarbinol, divinyl carbinol, acmc-20ap1x, 1,4-pentadien-3-ol, contains hydroquinone as stabilizer | |
OC(C=C)C=C | |
approximately 0.4% hydroquinone | |
70204 | |
98% | |
1, 4-Pentadien-3-ol, Stabilized |
97.5 | |
Colorless | |
115.0°C | |
Authentic | |
C5H8O | |
1.4440 to 1.4460 (20°C, 589nm) | |
0.865 | |
ICMWSAALRSINTC-UHFFFAOYSA-N | |
penta-1,4-dien-3-ol | |
84.12 | |
84.12 | |
Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 213-080-9
- 1,4-Pentadien-3-ol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only