missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Pentadiene, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -148.0°C |
Color | Colorless |
Density | 0.6500g/mL |
Boiling Point | 26.0°C |
Flash Point | −20°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 1,4-Pentadiene
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 1
- H304-May be fatal if swallowed and enters airways.
- H224-Extremely flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'