Learn More
1,4-Pentadiene, 99%, Thermo Scientific Chemicals
£27.90 - £86.10
Chemical Identifiers
CAS | 591-93-5 |
---|---|
Molecular Formula | C5H8 |
Molecular Weight (g/mol) | 68.11 |
MDL Number | MFCD00008665 |
InChI Key | QYZLKGVUSQXAMU-UHFFFAOYSA-N |
Synonym | 1,4-pentadiene, unii-a5if861oq5, pentadiene-1,4, allylethylene, penta-1,4-dien-1-yl, penta-1,4-dien-3-yl, ch2=chch2ch=ch2, acmc-1at58 |
PubChem CID | 11587 |
IUPAC Name | penta-1,4-diene |
SMILES | C=CCC=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10483395
|
Thermo Scientific Acros
129860010 |
1 g | Glass bottle |
£27.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10376960
|
Thermo Scientific Acros
129860050 |
5 g | Glass bottle |
£86.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
591-93-5 | |
68.11 | |
QYZLKGVUSQXAMU-UHFFFAOYSA-N | |
11587 | |
C=CCC=C |
C5H8 | |
MFCD00008665 | |
1,4-pentadiene, unii-a5if861oq5, pentadiene-1,4, allylethylene, penta-1,4-dien-1-yl, penta-1,4-dien-3-yl, ch2=chch2ch=ch2, acmc-1at58 | |
penta-1,4-diene |
Specifications
591-93-5 | |
100.0 | |
Colorless | |
26.0°C | |
Authentic | |
C5H8 | |
1.3884 to 1.3904 | |
01, 251 | |
0.65 | |
QYZLKGVUSQXAMU-UHFFFAOYSA-N | |
penta-1,4-diene | |
11587 | |
99% | |
1,4-Pentadiene, 99% |
98.5 | |
-148.0°C | |
0.6500g/mL | |
−20°C | |
99% | |
H2C=CHCH2CH=CH2 | |
MFCD00008665 | |
01,783 | |
1,4-pentadiene, unii-a5if861oq5, pentadiene-1,4, allylethylene, penta-1,4-dien-1-yl, penta-1,4-dien-3-yl, ch2=chch2ch=ch2, acmc-1at58 | |
C=CCC=C | |
68.11 | |
68.11 | |
Liquid |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfac
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-736-9
missing translation for 'tsca' : TSCA
- 1,4-Pentadiene
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H224-Extremely flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only