missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylenediacryloyl chloride, tech., Thermo Scientific™

Product Code. 15447937
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35288-49-4
Molecular Formula C12H8Cl2O2
Molecular Weight (g/mol) 255.094
MDL Number MFCD01109676
InChI Key WQZLRZFBCIUDFL-KQQUZDAGSA-N
Synonym 1,4-phenylenediacryloyl chloride, 3,3'-1,4-phenylene bis 2-propenoyl chloride, 3,3'-benzene-1,4-diylbisprop-2-enoyl chloride, 1 4-phenylenediacryloyl chloridetech, 3,3'-p-phenylene bis acrylic acid chloride, 1,4-phenylenediacryloyl chloride, technical grade, 2-propenoyl chloride, 3,3'-1,4-phenylene bis, 3,3'-p-phenylene bis e-acrylic acid chloride, 2e,2'e-3,3'-1,4-phenylene diacryloyl chloride, e-3-4-e-3-chloro-3-oxoprop-1-enyl phenyl prop-2-enoyl chloride
PubChem CID 5989690
IUPAC Name (E)-3-[4-[(E)-3-chloro-3-oxoprop-1-enyl]phenyl]prop-2-enoyl chloride
SMILES C1=CC(=CC=C1C=CC(=O)Cl)C=CC(=O)Cl

Specifications

Quantity 5 g
UN Number UN1759
Sensitivity Moisture sensitive
Formula Weight 255.1
Grade Technical
Chemical Name or Material 1,4-Phenylenediacryloyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'