missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Pyridyl)piperazine, ACROS Organics™
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Specifications
CAS | 1008-91-9 |
Packaging | Glass bottle |
Molecular Formula | C9H13N3 |
Quantity | 5 g |
Synonym | 1-4-pyridyl piperazine, 1-pyridin-4-yl piperazine, 1-pyridin-4-yl-piperazine, 4-piperazinopyridine, 4-pyridylpiperazine, 1-4-pyridyl-piperazine, 4-1-piperazino pyridine, 1-4-pyridinyl piperazine, piperazine, 1-4-pyridinyl |
InChI Key | OQZBAQXTXNIPRA-UHFFFAOYSA-N |
SMILES | C1CN(CCN1)C2=CC=NC=C2 |
IUPAC Name | 1-pyridin-4-ylpiperazine |
Molecular Weight (g/mol) | 163.224 |
PubChem CID | 70517 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- 1-(4-Pyridyl)piperazine
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'