missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Trifluoromethyl)-2-pyrimidinyl]-4-piperidinecarboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10640725
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 465514-39-0
Molecular Formula C11H12F3N3O2
Molecular Weight (g/mol) 275.23
MDL Number MFCD02091615
InChI Key QYAIISDWPUEPHG-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-4-trifluoromethyl pyrimidin-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidine carboxylic acid, 1-4-trifluoromethyl pyrimid-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-pyrimidin-2-yl-4-piperidinecarboxylic acid
PubChem CID 2777798
IUPAC Name 1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
SMILES OC(=O)C1CCN(CC1)C1=NC=CC(=N1)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 275.23
Percent Purity 97%
Chemical Name or Material 1-[4-(Trifluoromethyl)-2-pyrimidinyl]-4-piperidinecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'