missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 246.23 |
Percent Purity | 95% |
Chemical Name or Material | 1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1, 4-diazepane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane, 95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction