missing translation for 'onlineSavingsMsg'
Learn More

1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 11316965
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 11316965

Brand: Thermo Scientific Maybridge TL00212EB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17610-21-8
Molecular Formula C16H22O
Molecular Weight (g/mol) 230.351
InChI Key IHUSZOMIBSDQTB-UHFFFAOYSA-N
Synonym 1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one, 1,1,4,4-tetramethyl-6-acetyltetralin, 1-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethanone, 1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-naphthalen-2-yl-ethanone, 6-acetyl-1,2,3,4-tetrahydro-1,1,4,4-tetramethylnaphthalene, 2-acetyl-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene, 6-acetyl-1,1,4,4-tetramethyltetralin, 6-acetyl-1,1,4,4-tetra-methyl-tetralin, 1,1,4,4-tetramethyl-1,2,3,4-tetrahydro-6-acetylnaphthalene
PubChem CID 2747562
IUPAC Name 1-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)C(CCC2(C)C)(C)C

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 25 g
Formula Weight 230.35
Percent Purity 95%
Chemical Name or Material 1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.