missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione, Tech., Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 50 g |
Formula Weight | 270.24 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'