Learn More
Chemical Identifiers
CAS | 462-94-2 |
---|---|
Molecular Formula | C5H14N2 |
Molecular Weight (g/mol) | 102.18 |
MDL Number | MFCD00008239 |
InChI Key | VHRGRCVQAFMJIZ-UHFFFAOYSA-N |
Synonym | 1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p |
PubChem CID | 273 |
ChEBI | CHEBI:18127 |
IUPAC Name | pentane-1,5-diamine |
SMILES | NCCCCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10675142
|
Thermo Scientific Acros
112320050 |
5 mL | Glass bottle |
£96.38
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10685142
|
Thermo Scientific Acros
112320250 |
25 mL | Glass bottle |
£312.56
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
462-94-2 | |
102.18 | |
VHRGRCVQAFMJIZ-UHFFFAOYSA-N | |
273 | |
pentane-1,5-diamine |
C5H14N2 | |
MFCD00008239 | |
1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p | |
CHEBI:18127 | |
NCCCCCN |
Specifications
462-94-2 | |
100.0 | |
Colorless to Yellow | |
178°C to 180°C | |
Authentic | |
C5H14N2 | |
1.4570 to 1.463 | |
04, 266 | |
0.87 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ether | |
NCCCCCN | |
102.18 | |
CHEBI:18127 | |
98% | |
1, 5-Diaminopentane, 98% |
97.5 | |
9°C | |
0.8700g/mL | |
62°C | |
97.5% min. (GC) | |
H2N(CH2)5NH2 | |
MFCD00008239 | |
15, 1614 | |
1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p | |
VHRGRCVQAFMJIZ-UHFFFAOYSA-N | |
pentane-1,5-diamine | |
273 | |
102.18 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 207-329-0
missing translation for 'rtecsNumber' : SA0200000
missing translation for 'tsca' : TSCA
- 1,5-Diaminopentane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Pentamethylenediamine
RUO – Research Use Only