Learn More
(1,5-Dimethyl-1H-pyrazol-3-yl)methanol, 97%, Thermo Scientific™
£39.15 - £80.55
Identificatori chimici
CAS | 153912-60-8 |
---|---|
Molecular Formula | C6H10N2O |
Molecular Weight (g/mol) | 126.159 |
MDL Number | MFCD03659702 |
InChI Key | XJBMHIHXRARJJS-UHFFFAOYSA-N |
Synonym | 1,5-dimethyl-1h-pyrazol-3-yl methanol, 1,5-dimethyl-1h-pyrazol-3-yl-methanol, 1,5-dimethylpyrazol-3-yl methanol, 3-hydroxymethyl-1,5-dimethyl-1h-pyrazole, 1h-pyrazole-3-methanol,1,5-dimethyl, 1,5-dimethyl-3-hydroxymethyl-1h-pyrazole, 1,5-dimethyl-3-hydroxymethyl pyrazole, akos pao-0060, acmc-1cisl, 1,5-dimethylpyrazole-3-methanol |
PubChem CID | 2776366 |
IUPAC Name | (1,5-dimethylpyrazol-3-yl)methanol |
SMILES | CC1=CC(=NN1C)CO |
Codice del prodotto | Marca | Quantity | Packaging | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Packaging | Prezzo | Quantità e disponibilità | ||||
10190912
|
Thermo Scientific Maybridge
CC12609CB |
250 mg | Amber Glass Bottle |
£39.15
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10652243
|
Thermo Scientific Maybridge
CC12609DA |
1 g | Amber Glass Bottle |
£80.55
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
Identificatori chimici
153912-60-8 | |
126.159 | |
XJBMHIHXRARJJS-UHFFFAOYSA-N | |
2776366 | |
CC1=CC(=NN1C)CO |
C6H10N2O | |
MFCD03659702 | |
1,5-dimethyl-1h-pyrazol-3-yl methanol, 1,5-dimethyl-1h-pyrazol-3-yl-methanol, 1,5-dimethylpyrazol-3-yl methanol, 3-hydroxymethyl-1,5-dimethyl-1h-pyrazole, 1h-pyrazole-3-methanol,1,5-dimethyl, 1,5-dimethyl-3-hydroxymethyl-1h-pyrazole, 1,5-dimethyl-3-hydroxymethyl pyrazole, akos pao-0060, acmc-1cisl, 1,5-dimethylpyrazole-3-methanol | |
(1,5-dimethylpyrazol-3-yl)methanol |
Specifica
153912-60-8 | |
100.0 | |
C6H10N2O | |
1,5-dimethyl-1h-pyrazol-3-yl methanol, 1,5-dimethyl-1h-pyrazol-3-yl-methanol, 1,5-dimethylpyrazol-3-yl methanol, 3-hydroxymethyl-1,5-dimethyl-1h-pyrazole, 1h-pyrazole-3-methanol,1,5-dimethyl, 1,5-dimethyl-3-hydroxymethyl-1h-pyrazole, 1,5-dimethyl-3-hydroxymethyl pyrazole, akos pao-0060, acmc-1cisl, 1,5-dimethylpyrazole-3-methanol | |
CC1=CC(=NN1C)CO | |
126.159 | |
126.16 | |
(1, 5-Dimethyl-1H-pyrazol-3-yl)methanol |
96.5 | |
97% | |
MFCD03659702 | |
XJBMHIHXRARJJS-UHFFFAOYSA-N | |
(1,5-dimethylpyrazol-3-yl)methanol | |
2776366 | |
97% |
Sicurezza e movimentazione
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.