missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1,5-Diphenylcarbazide is used in the colorimetric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 170°C to 176°C |
Color | White to Cream |
Quantity | 5 g |
Beilstein | 752039 |
Sensitivity | Air and light sensitive |
Merck Index | 14,3321 |
Solubility Information | Soluble in glacial acetic acid,alcohol and acetone; Slightly soluble in water. |
Formula Weight | 242.28 |
Grade | ACS |
Physical Form | Crystalline |
Show More |
Safety and Handling
Product Identifier
- 1,5-Diphenylcarbazide
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction