missing translation for 'onlineSavingsMsg'
Learn More
1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals Product Code.: 11382917

1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals

Product Code. 11382917
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11382917

Brand: Thermo Scientific Alfa Aesar L16185.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-95-5
Molecular Formula C6H10N4
Molecular Weight (g/mol) 138.174
MDL Number MFCD00005939
InChI Key CWRVKFFCRWGWCS-UHFFFAOYSA-N
Synonym pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazoleShow MoreShow Less
PubChem CID 5917
ChEBI CHEBI:34910
IUPAC Name 6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
SMILES C1CCC2=NN=NN2CC1

Specifications

Melting Point 59°C to 61°C
Boiling Point 194°C (12 mmHg)
Quantity 25 g
UN Number UN2811
Beilstein 135492
Sensitivity Hygroscopic
Merck Index 14,7141
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 1,5-Pentamethylene-1H-tetrazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249792 Certificate TypeCertificate of Analysis Date06/03/2024 Catalog NumberL16185.06,L16185.14
Lot Number10249655 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog NumberL16185.06,L16185.14
Lot Number10248590 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog NumberL16185.06,L16185.14
Lot Number10238982 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL16185.06,L16185.14
Lot Number10239332 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL16185.06,L16185.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.