missing translation for 'onlineSavingsMsg'
Learn More

1-[5-(Trifluoromethyl)-2-pyridyl]-1,4-diazepane,95+%, Thermo Scientific™

£18.40 - £71.30

Chemical Identifiers

CAS 306934-70-3
Molecular Formula C11H14F3N3
Molecular Weight (g/mol) 245.25
MDL Number MFCD00661783
InChI Key IBMSHQVIAUTKDL-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazepane, 1-5-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4 diazepane, 1h-1,4-diazepine,hexahydro-1-5-trifluoromethyl-2-pyridinyl, 1-5-trifluoromethyl-pyridin-2-yl 1,4-diazepane, 1-5-trifluoromethyl-2-pyridinyl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine
PubChem CID 2760779
IUPAC Name 1-[5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
SMILES FC(F)(F)C1=CN=C(C=C1)N1CCCNCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10308483
View Documents
Thermo Scientific Maybridge
AW00291DA
1 g Amber Glass Bottle
£18.40
1g
Please to purchase this item. Need a web account? Register with us today!
10680656
View Documents
Thermo Scientific Maybridge
AW00291DE
5 g Amber Glass Bottle
£71.30
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

306934-70-3
245.25
IBMSHQVIAUTKDL-UHFFFAOYSA-N
2760779
FC(F)(F)C1=CN=C(C=C1)N1CCCNCC1
C11H14F3N3
MFCD00661783
1-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazepane, 1-5-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4 diazepane, 1h-1,4-diazepine,hexahydro-1-5-trifluoromethyl-2-pyridinyl, 1-5-trifluoromethyl-pyridin-2-yl 1,4-diazepane, 1-5-trifluoromethyl-2-pyridinyl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine
1-[5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane

Specifications

306934-70-3
C11H14F3N3
1-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazepane, 1-5-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4 diazepane, 1h-1,4-diazepine,hexahydro-1-5-trifluoromethyl-2-pyridinyl, 1-5-trifluoromethyl-pyridin-2-yl 1,4-diazepane, 1-5-trifluoromethyl-2-pyridinyl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine
FC(F)(F)C1=CN=C(C=C1)N1CCCNCC1
245.25
245.25
1-[5-(Trifluoromethyl)-2-pyridyl]-1, 4-diazepane
97%
MFCD00661783
IBMSHQVIAUTKDL-UHFFFAOYSA-N
1-[5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
2760779
97%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications