Learn More
1,6-Anhydro-beta-D-glucopyranose, 99%, Thermo Scientific Chemicals
Description
1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.
Solubility
Soluble in Ethanol and Water.
Notes
Store at -20°C. Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 180°C to 184°C |
Quantity | 0.25 g |
Beilstein | 80998 |
Solubility Information | Soluble in Ethanol and Water. |
Formula Weight | 162.14 |
Percent Purity | 99% |
Chemical Name or Material | 1,6-Anhydro-beta-D-glucopyranose |
RUO – Research Use Only