missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 1,6-Diiodoperfluorohexane, 97%
£140.00
Specifications
Chemical Name or Material | 1,6-Diiodoperfluorohexane |
---|---|
CAS | 375-80-4 |
Melting Point | 25°C |
Density | 2.357 |
Boiling Point | 115°C (100 mmHg) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11399719
|
Thermo Scientific Chemicals
B21654.06 |
5 g |
£140.00
5g |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
It is employed in the synthesis and characterization of fluorinated telomers containing vinylidene fluoride and hexafluoropropene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed in the synthesis and characterization of fluorinated telomers containing vinylidene fluoride and hexafluoropropene.
Notes
Light sensitive. Protect from light. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
1,6-Diiodoperfluorohexane | |
25°C | |
115°C (100 mmHg) | |
1.398 | |
1822186 | |
JOQDDLBOAIKFQX-UHFFFAOYSA-N | |
1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane | |
78994 | |
97% |
375-80-4 | |
2.357 | |
C6F12I2 | |
MFCD00042264 | |
Light sensitive | |
FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I | |
553.86 | |
553.86 |
RUO – Research Use Only