Learn More
1,6-Diisocyanatohexane, 98+%, Thermo Scientific Chemicals
£33.60 - £111.00
Chemical Identifiers
CAS | 822-06-0 |
---|---|
Molecular Formula | C8H12N2O2 |
Molecular Weight (g/mol) | 168.20 |
MDL Number | MFCD00002047 |
InChI Key | RRAMGCGOFNQTLD-UHFFFAOYSA-N |
Synonym | hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester |
PubChem CID | 13192 |
ChEBI | CHEBI:53578 |
IUPAC Name | 1,6-diisocyanatohexane |
SMILES | O=C=NCCCCCCN=C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11320448
|
Thermo Scientific Alfa Aesar
043803.18 |
50 g |
£33.60
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11330448
|
Thermo Scientific Alfa Aesar
043803.30 |
250 g |
£111.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,6-Diisocyanatohexane is used as a polymerizing agent in polyurethane paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
822-06-0 | |
168.20 | |
RRAMGCGOFNQTLD-UHFFFAOYSA-N | |
13192 | |
1,6-diisocyanatohexane |
C8H12N2O2 | |
MFCD00002047 | |
hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester | |
CHEBI:53578 | |
O=C=NCCCCCCN=C=O |
Specifications
822-06-0 | |
1.05 g/mL | |
130°C (266°F) | |
C8H12N2O2 | |
MFCD00002047 | |
956709 | |
hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester | |
RRAMGCGOFNQTLD-UHFFFAOYSA-N | |
1,6-diisocyanatohexane | |
13192 | |
168.19 | |
Liquid |
-67°C | |
255°C | |
Pungent | |
1.452 | |
UN2281 | |
Moisture sensitive | |
Reacts with water. | |
O=C=NCCCCCCN=C=O | |
168.20 | |
CHEBI:53578 | |
≥98% | |
1,6-Diisocyanatohexane |
Safety and Handling
GHS H Statement
H331-H334-H315-H319-H317-H335-H204
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fire or projection hazard.
P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302-H314-H317-H330-H334-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: HEXAMETHYLENE DIISOCYANATE
missing translation for 'einecsNumber' : 212-485-8
missing translation for 'rtecsNumber' : MO1740000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only