missing translation for 'onlineSavingsMsg'
Learn More
1,6-Diisocyanatohexane, 99+%, Thermo Scientific Chemicals Product Code.: 10336130

1,6-Diisocyanatohexane, 99+%, Thermo Scientific Chemicals

Product Code. 10336130 Shop All Thermo Scientific Products
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10336130

Brand: Thermo Scientific 115190250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-06-0
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00002047
InChI Key RRAMGCGOFNQTLD-UHFFFAOYSA-N
Synonym hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene esterShow MoreShow Less
PubChem CID 13192
ChEBI CHEBI:53578
IUPAC Name 1,6-diisocyanatohexane
SMILES O=C=NCCCCCCN=C=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -67°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 255.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula OCN(CH2)6NCO
Refractive Index 1.4510 to 1.4530
Quantity 25 g
Beilstein 04, II, 711
Specific Gravity 1.04
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in benzene,acetone,toluene,,ethyl acetate,chlorobenzene,very soluble in ether
Formula Weight 168.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1, 6-Diisocyanatohexane, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463211 Certificate TypeCertificate of Analysis Date10/06/2024 Catalog Number115190010,115191000,115190250
Lot NumberA0437870 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number115190010,115191000,115190250
Lot NumberA0399204 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number115190010,115191000,115190250
Lot NumberA0413874 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number115190010,115191000,115190250
Lot NumberA0404284 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number115190010,115191000,115190250
compliance-icons compliance-icons
Product Identifier
  • 1,6-Diisocyanatohexane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,6-Diisocyanatohexane, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.