missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediamine, 99.5+%, Thermo Scientific Chemicals

Product Code. 10677652
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 124-09-4
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
InChI Key NAQMVNRVTILPCV-UHFFFAOYSA-N
Synonym 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
PubChem CID 16402
ChEBI CHEBI:39618
IUPAC Name hexane-1,6-diamine
SMILES C(CCCN)CCN

Specifica

Melting Point 38.0°C to 41.0°C
Color White
Boiling Point 204.0°C to 205.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Plastic bottle
Linear Formula H2N(CH2)6NH2
Quantity 1 kg
Beilstein 04,269
Merck Index 15,4727
Solubility Information Solubility in water: 490g/L (20°C). Other solubilities: soluble in ethanol, benzene, diethyl ether
Formula Weight 116.21
Percent Purity 99.5+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 1, 6-Hexanediamine
Vedi altri risultati Mostra meno risultati
compliance-icons compliance-icons
Identificativo prodotto
  • 1,6-Hexanediamine
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informazioni supplementari
  • MIXTURE LIST-Contains : 1,6-Hexanediamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'