missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
1,6-Hexanediol diacrylate is used as a functional monomer for polymers. It acts as a cross linking agent between the molecular chains of polymers. Further, it is used in adhesives, sealants, alkyd coatings, elastomers, photopolymers, and inks for improved adhesion, hardness, abrasion and heat resistance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.010 g/mL |
Flash Point | >110°C (>230°F) |
Quantity | 50 g |
Sensitivity | Hygroscopic |
Solubility Information | Slightly miscible with water. |
Formula Weight | 226.28 |
Percent Purity | 99% |
Physical Form | Liquid |
Assay | (reactive esters) |
Chemical Name or Material | 1,6-Hexanediol diacrylate, Stabilized with 90ppm hydroquinone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'