missing translation for 'onlineSavingsMsg'
Learn More
1,8-Cineole, 99%, Thermo Scientific Chemicals Product Code.: 10605532

1,8-Cineole, 99%, Thermo Scientific Chemicals

Product Code. 10605532
100 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10605532

Brand: Thermo Scientific Acros 110341000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 470-82-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167977
InChI Key WEEGYLXZBRQIMU-UHFFFAOYSA-N
Synonym eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
PubChem CID 2758
IUPAC Name 2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane
SMILES CC12CCC(CC1)C(C)(C)O2

Specifications

Melting Point 1.5°C
Color Colorless to Yellow
Density 0.9225g/mL
Boiling Point 176°C to 177°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4550 to 1.46
Quantity 100 g
Beilstein 17,II,32
Merck Index 15, 3938
Specific Gravity 0.9225
Specific Rotation -0.3° to +0.3° (20°C, 589nm) (neat)
Solubility Information (in ethanol (70% v/v)) 1:2
Formula Weight 154.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,8-Cineole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,8-Cineole
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : 1,8-Cineol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,8-Cineole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.