Learn More
Chemical Identifiers
CAS | 373-44-4 |
---|---|
Molecular Formula | C8H22Cl2N2 |
Molecular Weight (g/mol) | 217.18 |
MDL Number | MFCD00008248 |
InChI Key | ZFLWZOGXFQNIMT-UHFFFAOYSA-N |
Synonym | 1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392 |
PubChem CID | 24250 |
ChEBI | CHEBI:73112 |
SMILES | [Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10664952
|
Thermo Scientific Acros
112310250 |
25 g | Glass bottle |
£26.63
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10122810
|
Thermo Scientific Acros
112311000 |
100 g | Glass bottle |
£70.55
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10783455
|
Thermo Scientific Acros
112315000 |
500 g | Glass bottle |
£226.13
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
373-44-4 | |
217.18 | |
ZFLWZOGXFQNIMT-UHFFFAOYSA-N | |
24250 | |
[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+] |
C8H22Cl2N2 | |
MFCD00008248 | |
1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392 | |
CHEBI:73112 |
Specifications
373-44-4 | |
100.0 | |
White to Yellow | |
225.0°C to 226.0°C | |
Authentic | |
C8H22Cl2N2 | |
MFCD00008248 | |
1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392 | |
ZFLWZOGXFQNIMT-UHFFFAOYSA-N | |
octane-1,8-bis(aminium) dichloride | |
24250 | |
144.26 | |
Solidified Crystalline Mass |
97.5 | |
50.0°C to 54.0°C | |
165g/mL | |
115°C | |
97.5% min. (GC) | |
H2N(CH2)8NH2 | |
04, 271 | |
Solubility in water: 575g/L (20°C). | |
[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+] | |
217.18 | |
CHEBI:73112 | |
98% | |
1, 8-Diaminooctane |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physician.
Wear
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-764-3
- 1,8-Diaminooctane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only