Learn More
1,8-Diaminooctane, 98%, Thermo Scientific Chemicals
£31.76 - £79.29
Chemical Identifiers
CAS | 373-44-4 |
---|---|
Molecular Formula | C8H22Cl2N2 |
Molecular Weight (g/mol) | 217.18 |
MDL Number | MFCD00008248 |
InChI Key | ZFLWZOGXFQNIMT-UHFFFAOYSA-N |
Synonym | 1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392 |
PubChem CID | 24250 |
ChEBI | CHEBI:73112 |
IUPAC Name | octane-1,8-diamine |
SMILES | [Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11426736
|
Thermo Scientific Alfa Aesar
B23885.14 |
25 g |
£31.76
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11436736
|
Thermo Scientific Alfa Aesar
B23885.22 |
100 g |
£79.29
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,8-Diaminooctane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water 575g/L.
Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, carbon dioxide, air, water/ moisture.
Chemical Identifiers
373-44-4 | |
217.18 | |
ZFLWZOGXFQNIMT-UHFFFAOYSA-N | |
24250 | |
octane-1,8-diamine |
C8H22Cl2N2 | |
MFCD00008248 | |
1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392 | |
CHEBI:73112 | |
[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+] |
Specifications
373-44-4 | |
11.7 | |
106°C (223°F) | |
C8H22Cl2N2 | |
UN3259 | |
Air sensitive; Hygroscopic | |
Soluble in water 575g/L. | |
[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+] | |
217.18 | |
CHEBI:73112 | |
98% |
50°C to 54°C | |
225°C to 226°C | |
Amine-like | |
MFCD00008248 | |
1735426 | |
1,8-diaminooctane, 1,8-octanediamine, octamethylenediamine, 1,8-octylenediamine, 1,8-octamethylenediamine, diaminooctane, octane 1,8-diamine, unii-53a6694pie, alpha,omega-diaminooctane, chembl29392 | |
ZFLWZOGXFQNIMT-UHFFFAOYSA-N | |
octane-1,8-diamine | |
24250 | |
144.26 | |
1,8-Diaminooctane |
Safety and Handling
GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302-H314-H317-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 206-764-3
missing translation for 'rtecsNumber' : RG8841500
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 1,8-Diaminooctane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301-IF SWALLOWED:.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Octamethylenediamine
RUO – Research Use Only