missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%, Thermo Scientific Chemicals

Product Code. 10662023
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6674-22-2
MDL Number MFCD00006930
InChI Key GQHTUMJGOHRCHB-UHFFFAOYSA-N
Synonym 1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
PubChem CID 81184
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
SMILES C1CCC2=NCCCN2CC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 1.0190g/mL
Boiling Point 261.0°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C9H16N2
Refractive Index 1.5210 to 1.5230
Quantity 500 g
Beilstein 23, V,5, 271
Fieser 02,101; 04,119; 05,177; 06,158; 07,87; 11,155; 13,91; 14,109; 15,109; 16,105; 17,99
Merck Index 15, 2996
Specific Gravity 1.019
Solubility Information Solubility in water: soluble.
Molecular Weight (g/mol) 152.24
Viscosity 11.76 mPa.s (20°C)
Formula Weight 152.24
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]undec-7-ene, ≥98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,8-Diazabicyclo[5.4.0]undec-7-ene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,8-Diazabicyclo[5.4.0]undec-7-ene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.