missing translation for 'onlineSavingsMsg'
Learn More

1,8-Diazafluoren-9-one, Thermo Scientific Chemicals

Product Code. 15730909
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54078-29-4
Molecular Formula C11H6N2O
Molecular Weight (g/mol) 182.18
InChI Key FOSUVSBKUIWVKI-UHFFFAOYSA-N
Synonym 9h-cyclopenta 1,2-b:4,3-b' dipyridin-9-one, 1,8-diazafluoren-9-one, 1,8-diazafluoran-9-one, 9h-pyrido 3',2':3,4 cyclopenta 1,2-b pyridin-9-one, 6,10-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaen-8-one, 1.8-diazafluoren-9-one, 9h-1,8-diazafluoren-9-one, fosuvsbkuiwvki-uhfffaoysa, pyridino 2',3'-1,5 cyclopenta 3,4-b pyridin-9-one, 6,10-diazatricyclo 7.4.0.0^ 2,7 trideca-1 9 ,2,4,6,10,12-hexaen-8-one
PubChem CID 725961
SMILES C1=CC2=C(C(=O)C3=C2C=CC=N3)N=C1

Specifications

Melting Point 229.0°C to 233.0°C
Quantity 1 g
Formula Weight 182.18
Chemical Name or Material 1,8-Diazafluoren-9-one
compliance-icons
Product Identifier
  • 1,8-Diazafluoren-9-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,8-Diazafluoren-9-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'