missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dibromooctane, 98%, Thermo Scientific Chemicals

Product Code. 10326370
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-32-0
Molecular Formula C8H16Br2
Molecular Weight (g/mol) 272.01
InChI Key DKEGCUDAFWNSSO-UHFFFAOYSA-N
Synonym octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r
PubChem CID 78310
IUPAC Name 1,8-dibromooctane
SMILES C(CCCCBr)CCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 15.0°C to 16.0°C
Color Yellow
Density 1.4600g/mL
Boiling Point 270.0°C to 272.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Br(CH2)8Br
Refractive Index 1.4981 to 1.5001
Quantity 10 mL
Beilstein 01,16
Specific Gravity 1.46
Solubility Information Solubility in water: immiscible
Formula Weight 272.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 8-Dibromooctane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'