missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Naphthalic anhydride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
1,8-Naphthalic anhydride is used as a seed protectant and to prepare dyestuffs and optical brighteners. It is used in the preparation of napthalimide analogues, which finds application as anticancer agents. Further, it is used as fluorescent brightening agent for polymeric materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,8-Naphthalic anhydride is used as a seed protectant and to prepare dyestuffs and optical brighteners. It is used in the preparation of napthalimide analogues, which finds application as anticancer agents. Further, it is used as fluorescent brightening agent for polymeric materials.
Solubility
Soluble in ether and alcohol. Slightly soluble in acetic acid. Insoluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
1,8-Naphthalic anhydride is used as a seed protectant and to prepare dyestuffs and optical brighteners. It is used in the preparation of napthalimide analogues, which finds application as anticancer agents. Further, it is used as fluorescent brightening agent for polymeric materials.
Solubility
Soluble in ether and alcohol. Slightly soluble in acetic acid. Insoluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spécification
Melting Point | 269°C to 273°C |
Quantity | 50 g |
Beilstein | 153190 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ether and alcohol. Slightly soluble in acetic acid. Insoluble in water. |
Formula Weight | 198.18 |
Percent Purity | 97% |
Chemical Name or Material | 1,8-Naphthalic anhydride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'