missing translation for 'onlineSavingsMsg'
Learn More

1,9-Decadiene, 97%, Thermo Scientific Chemicals

Product Code. 11337428
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is widely used in the manufacturers and suppliers of pharmaceuticals and intermediates in the preparation of fine chemicals. 1,9-decadiene is also used as a starting material in ADMET polymerizations .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1647-16-1
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00008670
InChI Key NLDGJRWPPOSWLC-UHFFFAOYSA-N
Synonym 1,9-decadiene, decadiene-1,9, unii-2kwz01g244, acmc-1c2oy, dsstox_cid_2159, dsstox_rid_76506, dsstox_gsid_22159, chembl31020
PubChem CID 15439
IUPAC Name deca-1,9-diene
SMILES C=CCCCCCCC=C

Specifications

Density 0.75 g/mL
Boiling Point 169°C
Flash Point 42°C (107°F)
Refractive Index 1.4325
Quantity 10 g
UN Number UN3295
Solubility Information It is soluble in organic solvents.
Formula Weight 138.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1,9-Decadiene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'