Learn More
1,9-Decadiene, 97%, Thermo Scientific Chemicals
£70.70 - £306.00
Chemical Identifiers
CAS | 1647-16-1 |
---|---|
Molecular Formula | C10H18 |
Molecular Weight (g/mol) | 138.254 |
MDL Number | MFCD00008670 |
InChI Key | NLDGJRWPPOSWLC-UHFFFAOYSA-N |
Synonym | 1,9-decadiene, decadiene-1,9, unii-2kwz01g244, acmc-1c2oy, dsstox_cid_2159, dsstox_rid_76506, dsstox_gsid_22159, chembl31020 |
PubChem CID | 15439 |
IUPAC Name | deca-1,9-diene |
SMILES | C=CCCCCCCC=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337428
|
Thermo Scientific Alfa Aesar
031207.09 |
10 g |
£70.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357428
|
Thermo Scientific Alfa Aesar
031207.18 |
50 g |
£306.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is widely used in the manufacturers and suppliers of pharmaceuticals and intermediates in the preparation of fine chemicals. 1,9-decadiene is also used as a starting material in ADMET polymerizations .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1647-16-1 | |
138.254 | |
NLDGJRWPPOSWLC-UHFFFAOYSA-N | |
15439 | |
C=CCCCCCCC=C |
C10H18 | |
MFCD00008670 | |
1,9-decadiene, decadiene-1,9, unii-2kwz01g244, acmc-1c2oy, dsstox_cid_2159, dsstox_rid_76506, dsstox_gsid_22159, chembl31020 | |
deca-1,9-diene |
Specifications
1647-16-1 | |
169°C | |
C10H18 | |
MFCD00008670 | |
1,9-decadiene, decadiene-1,9, unii-2kwz01g244, acmc-1c2oy, dsstox_cid_2159, dsstox_rid_76506, dsstox_gsid_22159, chembl31020 | |
C=CCCCCCCC=C | |
138.254 | |
138.25 | |
Liquid |
0.75g/mL | |
42° (107°F) | |
1.4325 | |
UN3295 | |
NLDGJRWPPOSWLC-UHFFFAOYSA-N | |
deca-1,9-diene | |
15439 | |
97% | |
1,9-Decadiene |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only