missing translation for 'onlineSavingsMsg'
Learn More

1-Acetoxy-2-methoxybenzene, 98%

Product Code. 11409716
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11409716

Brand: Thermo Scientific Alfa Aesar B20242.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15212-03-0 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15212-03-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00017221
InChI Key BHJHPYFAYGAPLS-UHFFFAOYSA-N
Synonym guaiacol acetate, 1-acetoxy-2-methoxybenzene, guaiacyl acetate, o-acetoxyanisole, o-methoxyphenyl acetate, o-anisyl acetate, acetyl guaiacol, phenol, 2-methoxy-, acetate, o-acetylguaiacol, eucol
PubChem CID 61155
ChEBI CHEBI:86645
IUPAC Name (2-methoxyphenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1OC

Specifications

Quantity 100 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 1-Acetoxy-2-methoxybenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.