missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Acetyl-3-indolecarboxaldehyde, 98%, Thermo Scientific™
£19.10 - £58.80
Chemical Identifiers
CAS | 22948-94-3 |
---|---|
Molecular Formula | C11H9NO2 |
Molecular Weight (g/mol) | 187.2 |
MDL Number | MFCD00039691 |
InChI Key | LCJLFGSKHBDOAY-UHFFFAOYSA-N |
Synonym | 1-acetyl-1h-indole-3-carbaldehyde, n-acetylindole-3-carboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1-acetyl-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, 1-acetyl-3-formylindole, zlchem 389, pubchem7227, n-acetylindole-3-aldehyde, acmc-1crl1 |
PubChem CID | 89915 |
IUPAC Name | 1-acetylindole-3-carbaldehyde |
SMILES | CC(=O)N1C=C(C2=CC=CC=C21)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10215154
|
Thermo Scientific Acros
399700010 |
1 g | |||||||
10326494
|
Thermo Scientific Acros
399700050 |
5 g |
£58.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
22948-94-3 | |
187.2 | |
LCJLFGSKHBDOAY-UHFFFAOYSA-N | |
89915 | |
CC(=O)N1C=C(C2=CC=CC=C21)C=O |
C11H9NO2 | |
MFCD00039691 | |
1-acetyl-1h-indole-3-carbaldehyde, n-acetylindole-3-carboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1-acetyl-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, 1-acetyl-3-formylindole, zlchem 389, pubchem7227, n-acetylindole-3-aldehyde, acmc-1crl1 | |
1-acetylindole-3-carbaldehyde |
Specifications
22948-94-3 | |
97.5% min. (GC) | |
C11H9NO2 | |
1-acetyl-1h-indole-3-carbaldehyde, n-acetylindole-3-carboxaldehyde, 1-acetylindole-3-carboxaldehyde, 1-acetyl-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 1-acetyl, 1-acetyl-3-formylindole, zlchem 389, pubchem7227, n-acetylindole-3-aldehyde, acmc-1crl1 | |
CC(=O)N1C=C(C2=CC=CC=C21)C=O | |
187.2 | |
187.2 | |
1-Acetyl-3-indolecarboxaldehyde |
165°C | |
Glass Bottle | |
MFCD00039691 | |
LCJLFGSKHBDOAY-UHFFFAOYSA-N | |
1-acetylindole-3-carbaldehyde | |
89915 | |
98% |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 245-347-0
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Acetyl-3-indolecarboxaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only